Få mere at vide
Ethoprophos (~90%), TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Ethoprophos |
| CAS | 13194-48-4 |
| Molekylær formel | C8 H19 O2 P S2 |
| Mængde | 1 g |
| Synonym | Phosphorodithioic acid, O-ethyl S,S-dipropyl ester, Ethyl propyl phosphorodithioate ((EtO)(PrS)2PO) (7CI), ENT 27318, Ethoprop, Ethoprophos, Mobile, Mocap, Mocap 10G, Mocap 15G, Mocap 6EC, O-Ethyl S,S-dipropyl dithiophosphate, O-Ethyl S,S-dipropyl phosphorodithioate, Profos, Prophos, Prophos (ester), Rovokil, S,S-Dipropyl O-ethyl phosphorodithioate, VC 9-104 |
| InChI formel | InChI=1S/C8H19O2PS2/c1-4-7-12-11(9,10-6-3)13-8-5-2/h4-8H2,1-3H3 |
| SMIL | CCCSP(=O)(OCC)SCCC |
| IUPAC navn | 1-[ethoxy(propylsulfanyl)phosphoryl]sulfanylpropane |
| Molekylvægt (g/mol) | 242.34 |
| Formel vægt | 242.0564 |