Få mere at vide
Ethionamide Sulfoxide, TRC
Artikelnummer. 30135351
Skift visning
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135351 25 mg
30006545 10 mg
30012438 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30135351 Leverandør Toronto Research Chemicals Leverandørnr. TRCE89042325MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 536-28-7
Molekylær formel C8 H10 N2 O S
Molekylvægt (g/mol) 182.24
Synonym Ethionamide Sulphoxide
IUPAC navn (2-ethylpyridin-4-yl)-sulfinylmethanamine
SMIL CCc1cc(ccn1)C(=S=O)N
Tekniske data
Kemisk navn eller materiale Ethionamide Sulfoxide
Mængde 25 mg
InChI formel InChI=1S/C8H10N2OS/c1-2-7-5-6(3-4-10-7)8(9)12-11/h3-5H,2,9H2,1H3
Formel vægt 182.0514