Learn More
Memmert™ Ethernet Connection Cable for Computer Interface for UF55plus
Ethernet connection cable for use with Memmert Ovens.
Brand:  Memmert™ UF55plusE06189
216.37 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 17539495
  • 226.00 DKK / Each
Only null left
Estimated Shipment: 29-08-2025
Description
  • 5 m
  • Accessory
Specifications
Specifications
UF55plus Oven
USB Connection Cable
5 m
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Memmert™ Ethernet Connection Cable for Computer Interface for UF55plus > For Use With: UF55plus