Få mere at vide
Ethaverine Hydrochloride, Mikromol™
Artikelnummer. 17921616
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
17921616 100 mg
1 options
Artikelnummer. 17921616 Leverandør Mikromol Leverandørnr. MM1575.00
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Ethaverine Hydrochloride
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ethaverine Hydrochloride
CAS 985-13-7
Analyt- eller komponentnavne Ethaverine Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C24 H29 N O4 . Cl H
Certificeringer/compliance ISO 17034
Synonym Isoquinoline, 1-(3,4-diethoxybenzyl)-6,7-diethoxy-, hydrochloride (7CI,8CI), Isoquinoline, 1-[(3,4-diethoxyphenyl)methyl]-6,7-diethoxy-, hydrochloride (9CI), 6,7-Diethoxy-1-(3,4-diethoxybenzyl)isoquinoline hydrochloride, Barbonin hydrochloride, Diquinol, Ethabid, Ethaverine hydrochloride, Isovex, Laverin, Papetherine hydrochloride, Perparin, Perparin hydrochloride, Perparine hydrochloride, Perperine, Perperine hydrochloride, Ethylpapaverine
InChI formel InChI=1S/C24H29NO4.ClH/c1-5-26-21-10-9-17(14-22(21)27-6-2)13-20-19-16-24(29-8-4)23(28-7-3)15-18(19)11-12-25-20;/h9-12,14-16H,5-8,13H2,1-4H3;1H
SMIL Cl.CCOc1ccc(Cc2nccc3cc(OCC)c(OCC)cc23)cc1OCC
IUPAC navn 1-[(3,4-diethoxyphenyl)methyl]-6,7-diethoxyisoquinoline;hydrochloride
Molekylvægt (g/mol) 431.95
Mængde 100 mg
Formel vægt 431.1863 g/mol