Få mere at vide
Ethaverine Hydrochloride, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Ethaverine Hydrochloride |
| CAS | 985-13-7 |
| Analyt- eller komponentnavne | Ethaverine Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C24 H29 N O4 . Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Isoquinoline, 1-(3,4-diethoxybenzyl)-6,7-diethoxy-, hydrochloride (7CI,8CI), Isoquinoline, 1-[(3,4-diethoxyphenyl)methyl]-6,7-diethoxy-, hydrochloride (9CI), 6,7-Diethoxy-1-(3,4-diethoxybenzyl)isoquinoline hydrochloride, Barbonin hydrochloride, Diquinol, Ethabid, Ethaverine hydrochloride, Isovex, Laverin, Papetherine hydrochloride, Perparin, Perparin hydrochloride, Perparine hydrochloride, Perperine, Perperine hydrochloride, Ethylpapaverine |
| InChI formel | InChI=1S/C24H29NO4.ClH/c1-5-26-21-10-9-17(14-22(21)27-6-2)13-20-19-16-24(29-8-4)23(28-7-3)15-18(19)11-12-25-20;/h9-12,14-16H,5-8,13H2,1-4H3;1H |
| Løsningstype | Neat |
| SMIL | Cl.CCOc1ccc(Cc2nccc3cc(OCC)c(OCC)cc23)cc1OCC |
| Vis mere |