missing translation for 'onlineSavingsMsg'
Learn More

Ethanolamine hydrochloride, 99+%, Thermo Scientific Chemicals

Catalog Number p-126444
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
2002-24-6
C2H7NO·HCl
97.55
MFCD00012892
PMUNIMVZCACZBB-UHFFFAOYSA-N
2-aminoethanol hydrochloride, ethanolamine hydrochloride, ethanolamine hcl, mea hydrochloride, monoethanolamine hydrochloride, ethanolamine chloride, colamine hydrochloride, ethanol, 2-amino-, hydrochloride, ethanolamine-hcl, 2-hydroxyethylammonium chloride
74819
2-aminoethanol;hydrochloride
C(CO)N.Cl
2002-24-6
C2H7NO·HCl
97.55
MFCD00012892
PMUNIMVZCACZBB-UHFFFAOYSA-N
2-aminoethanol hydrochloride, ethanolamine hydrochloride, ethanolamine hcl, mea hydrochloride, monoethanolamine hydrochloride, ethanolamine chloride, colamine hydrochloride, ethanol, 2-amino-, hydrochloride, ethanolamine-hcl, 2-hydroxyethylammonium chloride
74819
2-aminoethanol;hydrochloride
C(CO)N.Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 84°C to 86°C
Color Beige or White
Infrared Spectrum Authentic
Assay Percent Range 99% min. (Argentometry)
Packaging Glass bottle
Linear Formula H2NCH2CH2OH·HCl
Quantity 250 g
Beilstein 04, 275
Merck Index 15, 3782
Solubility Information Solubility in water: soluble
Formula Weight 97.55
Percent Purity 99+%
Physical Form Agglomerating Crystals
Chemical Name or Material Ethanolamine hydrochloride, p.a.
Show More Show Less
compliance-icons
Product Identifier
  • Ethanolamine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'