missing translation for 'onlineSavingsMsg'
Learn More

Ethanol, denaturated with 1% MEK and 0.001% Bitrex, 96%, Acros Organics™

Brand:  Acros Organics 457900250

Additional Details : CAS Number : 64-17-5 Weight : 19.75000kg Transport : UN number : 1170 Chem class : 3 Pack group : II

Quantity 25L

 View more versions of this product

Product Code. 15402192

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +45 70 27 99 20 for assistance


Specifications

Chemical Identifiers

64-17-5
46.069
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alkohol, anhydrol, tecsol
CHEBI:16236
CCO
C2H6O
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
702
ethanol

Specifications

Ethanol, denaturated with 1% MEK and 0.001% Bitrex
C2H6O
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alkohol, anhydrol, tecsol
CCO
46.069
CHEBI:16236
96%
0.79
1.2 mPa.s (20°C)
-114.0°C
3734-33-6
C2H5OH
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethanol
702
46.06
0.7900g/mL
12°C
78.0°C
25L
Videos