Få mere at vide
Ethanol Absolut Ph.Eur., Solveco
Beskrivelse
A synthetic ethanol that is produced through a complex three-step process, resulting in double-distilled ethanol. This makes it an ideal product for applications requiring the highest quality ethanol. Synthetic ethanol is manufactured using selective synthetic chemistry, which eliminates risks associated with contaminants such as genetically modified organisms, pesticide residues, natural allergens, phthalates, and dioxins. With a purity of 99.9%, this ethanol meets the quality requirements of the European Pharmacopoeia and is available with a batch-specific Certificate of Analysis upon request.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Ethyl Alcohol |
| Mængde | 1 L |
| Emballage | HDPE Bottle |
| Kogepunkt | 78°C |
| Procent renhed | 99.90% |
Sikkerhed og håndtering
- Etanol 99,7%
- Fare
- Alvorlige øjenskader/øjenirritation Kategori 2
- Brandfarlig væske Kategori 2
- H225-Meget brandfarlig væske og damp.
- H319-Forårsager alvorlig øjenirritation.
- P210-Holdes væk fra varme, varme overflader, gnister, åben ild og andre antændelseskilder. Rygning forbudt.
- P280-Bær beskyttelseshandsker/beskyttelsestøj/øjenbeskyttelse/ ansigtsbeskyttelse.
- P337+P313-Ved vedvarende øjenirritation: Søg lægehjælp.
- P370+P378-Ved brand: Anvend til brandslukning.
- P403+P235-Opbevares på et godt ventileret sted. Opbevares køligt.
- P501b-Indholdet/beholderen bortskaffes i henhold til de lokale/regionale/nationale/internationale forskrifter.
- MIXTURE LIST-Contain: Ethanol