Få mere at vide
Ethanol Absolut +MEK, Solveco
Artikelnummer. 18220275
Skift visning
Mængde:
1 L
5 L
25 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18220275 5 L
18250275 25 L
11514149 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
Artikelnummer. 18220275 Leverandør Solveco Chemicals Leverandørnr. 1029
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
Fermenteret og rektificeret ethanol denatureret med methylethylketon. Alkoholindholdet når 99,9 vol - %.
Fermenteret og rektificeret ethanol denatureret med methylethylketon. Alkoholindholdet når 99,9 vol - %.
Kemiske identifikatorer
CAS 64-17-5
Molekylær formel C2H6O
Molekylvægt (g/mol) 46.07
InChI nøgle LFQSCWFLJHTTHZ-UHFFFAOYSA-N
IUPAC navn ethanol
SMIL CCO
Tekniske data
Kemisk navn eller materiale Ethyl Alcohol
Mængde 5 L
Emballage HDPE beholder
Kogepunkt 78°C
Procent renhed 99.90%
Grad Teknisk