missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Solveco Ethanol A 96%, ETAX
Meets the requirements of the European Pharmacoupen. Solveco Ethanol A 96%, ETAX is produced by the fermentation of barley. It is a spectrographically pure ethanol of very high purity (PA-quality).
352.00 DKK - 1313.11 DKK
Chemical Identifiers
CAS | 64-17-5 |
---|---|
Molecular Formula | C2H6O |
Molecular Weight (g/mol) | 46.069 |
InChI Key | LFQSCWFLJHTTHZ-UHFFFAOYSA-N |
Synonym | ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alkohol, anhydrol, tecsol |
PubChem CID | 702 |
ChEBI | CHEBI:16236 |
IUPAC Name | ethanol |
SMILES | CCO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12654613
|
Solveco
1063 |
12 x 1L |
1313.11 DKK
12 L Qty Price Break Available |
In stock, ready to ship: 2 | Please sign in to purchase this item. Need a web account? Register with us today! |
12667213
|
Solveco
1001 |
3 x 5L |
352.00 DKK
5 L |
Estimated Shipment 13-04-2021 |
Please sign in to purchase this item. Need a web account? Register with us today! |
Description
This product conforms and is tested for meeting the requirements of the European Pharmacoupen. This ethanol is a food grade product that can be delivered with batchknutna analysis certificates as well as GMO and Kosher certificates.
- Free from allergens and a natural product of agricultural origin (barley)
- Molecular formula: CH3CH2OH
- CAS: 64-17-5
- Reacts explosively with strong oxidizing agents and can react violently with metals to produce hydrogen
- May damage gaskets, painted surfaces, protective and sealing grease coatings, materials, rubber and some plastics
Chemical Identifiers
64-17-5 | |
46.069 | |
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alkohol, anhydrol, tecsol | |
CHEBI:16236 | |
CCO |
C2H6O | |
LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
702 | |
ethanol |
Specifications
<0.40 (UV 5cm, 240nm) | |
<10 ppm | |
<2 ppm | |
LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
ethanol | |
702 | |
46.07g/mol | |
0.8036–0.8109kg/L | |
Bottles, HDPE | |
25 ppm | |
Colorless |
64-17-5 | |
C2H6O | |
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alkohol, anhydrol, tecsol | |
CCO | |
46.069 | |
CHEBI:16236 | |
95.1–96.9% | |
<200 ppm | |
PA, Food Quality Grade | |
Store at 18–30°C for up to 2 years in sealed package. | |
12 x 1L |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Etanol 96%
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.