missing translation for 'onlineSavingsMsg'
Learn More

Ethanol A 96%, ETAX, Solveco

Catalog Number 12667213
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Meets the requirements of the European Pharmacoupen. Solveco Ethanol A 96%, ETAX is produced by the fermentation of barley. It is a spectrographically pure ethanol of very high purity (PA-quality).

This product conforms and is tested for meeting the requirements of the European Pharmacoupen. This ethanol is a food grade product that can be delivered with batchknutna analysis certificates as well as GMO and Kosher certificates.

  • Free from allergens and a natural product of agricultural origin (barley)
  • Molecular formula: CH3CH2OH
  • CAS: 64-17-5
  • Reacts explosively with strong oxidizing agents and can react violently with metals to produce hydrogen
  • May damage gaskets, painted surfaces, protective and sealing grease coatings, materials, rubber and some plastics
compliance-icons
Product Identifier
  • Etanol 96%
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'