missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanol A 96%, ETAX, Solveco
Click to view available options
Quantity:
12 x 1 L
Description
This product conforms and is tested for meeting the requirements of the European Pharmacoupen. This ethanol is a food grade product that can be delivered with batchknutna analysis certificates as well as GMO and Kosher certificates.
- Free from allergens and a natural product of agricultural origin (barley)
- Molecular formula: CH3CH2OH
- CAS: 64-17-5
- Reacts explosively with strong oxidizing agents and can react violently with metals to produce hydrogen
- May damage gaskets, painted surfaces, protective and sealing grease coatings, materials, rubber and some plastics
Specifications
Absorbance | <0.40 (UV 5cm, 240nm) |
CAS | 64-17-5 |
Color | Colorless |
Density | 0.8036–0.8109kg/L |
Quantity | 12 x 1 L |
Packaging | HDPE Bottle |
Residue after Evaporation | 25 ppm |
Molecular Formula | C2H6O |
Synonym | ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol |
InChI Key | LFQSCWFLJHTTHZ-UHFFFAOYSA-N |
Show More |
Safety and Handling
Product Identifier
- Etanol 96%
- Danger
- Flammable liquid Category 2
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'