missing translation for 'onlineSavingsMsg'
Learn More

Ethanol 99.5% Analytical Grade, Solveco

Catalog Number 12634800
Click to view available options
Quantity:
3 x 5 L
12 x 1 L
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
64-17-5
C2H6O
46.069
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
702
CHEBI:16236
ethanol
CCO
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
64-17-5
C2H6O
46.069
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
702
CHEBI:16236
ethanol
CCO

Well suited for routine laboratory and analysis. Solveco Ethanol 99.5% Analytical Grade is fermented and rectified ethanol with very high purity. This PA quality solution has a natural origin. Available in 5L storage bottles or 1L bottles that can be directly used with a spray nozzle.

Denatured Ethanol 99.5% is a clear, colorless, volatile liquid with a distinctive odor. It reacts explosively with strong oxidizing agents and can react violently with metals to produce hydrogen. It may damage gaskets, painted surfaces, protective and sealing grease coatings, materials, rubber and some plastics.

  • PA quality, analytical grade
  • Molecular formula: CH3CH2OH
  • CAS: 64-17-5
Absorbance ≤0.500 (UV 5 cm, 240 nm)
Color Colorless
Formula Weight 46.07g/mol
Percent Purity ≥99.8%
Density ≤0.790 g/cm3
Quantity 3 x 5 L
Packaging HDPE Bottle
compliance-icons
Product Identifier
  • Etanol 99,5%
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'