missing translation for 'onlineSavingsMsg'
Learn More

Ethanol ≥ 96% (v/v), BAKER ANALYZED™ ACS, J.T. Baker™

Catalog Number p-7220839
Click to view available options
Quantity:
1 L
2.5 L
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
64-17-5
C2H6O
46.07
MFCD00003568
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethanol
CCO
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
64-17-5
C2H6O
46.07
MFCD00003568
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethanol
CCO

Exceeds ACS Specifications

Melting Point -114°C
Color Max. 10 (APHA)
Assay Percent Range 95.1 to 96.9% (v/v)
Quantity 1 L
Residue after Evaporation max. 0.001%
Titratable Acid max. 0.0005 meq/g
Titratable Base max. 0.0002 meq/g
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'