Få mere at vide
Ethanol 96% +MEK, Solveco
Artikelnummer. 11524149
Skift visning
Mængde:
1 L
5 L
Pakningsstørrelse:
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
11524149 1 L -
12486981 5 L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 11524149 Leverandør Solveco Chemicals Leverandørnr. 1129
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Fermented ethanol with a concentration of 96 vol - %, denatured with methyl ethyl ketone. Technical grade.
Fermented ethanol denatured with methylethylketone. The ethanol has a natural origin and is part of the natural cycle. Solveco Ethanol 96% +MEK is used extensively in the engineering industry and in washing fluids, as well as a coolant in machining.
Kemiske identifikatorer
CAS 64-17-5
Molekylær formel C2H6O
Molekylvægt (g/mol) 46.07
InChI nøgle LFQSCWFLJHTTHZ-UHFFFAOYSA-N
IUPAC navn ethanol
SMIL CCO
Tekniske data
Kemisk navn eller materiale Ethyl Alcohol
Mængde 1 L
Emballage HDPE Bottle
Kogepunkt 78°C
Procent renhed 96%
Grad Technical