Få mere at vide
Ethanol 70% WFI 1L, Solveco
Beskrivelse
Sterile-filtered and sterile-controlled ethanol at a concentration of 70 vol - %, diluted with Water for Injection (WFI). The raw materials used in this ethanol comply with and are tested against the quality standards of the current European Pharmacopoeia. Solveco recommends Ethanol 70% WFI for cleaning and disinfection purposes. This product effectively prevents the spread of spores and inhibits bacterial growth. Additionally, it is also available with preinstalled triggers (art 1439) for enhanced convenience and ease of use.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Ethyl Alcohol |
| CAS | 64-17-5 |
| Mængde | 1 L |
| Fysisk form | Liquid |
| Procent renhed | 70% |