missing translation for 'onlineSavingsMsg'
Få mere at vide

Ethanol 70% +MEK, Solveco

Artikelnummer. 30281156
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
15 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30281156 15 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
Artikelnummer. 30281156 Leverandør Solveco Chemicals Leverandørnr. 1052

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Fermented ethanol with a concentration of 70 vol - %, denatured with methyl ethyl ketone. Technical grade.

Fermented ethanol with a concentration of 70 vol - %, denatured with methyl ethyl ketone. Technical grade. The ethanol has a natural origin and is part of the natural cycle.

Tekniske data

Kemisk navn eller materiale Ethyl Alcohol
CAS 64-17-5
Mængde 15 L
Emballage 5 L HDPE Bottle
Koncentration 70%
Kogepunkt 78°C
Molekylvægt (g/mol) 46.07 g/mol
Procent renhed 70%
Grad Technical
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.