missing translation for 'onlineSavingsMsg'
Få mere at vide

Ethanol 70% +IPA Ph.Eur., Solveco

Artikelnummer. 12406991
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 L
25 L
Pakningsstørrelse:
25 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
12406991 25 L 25 L
18270275 5 L -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
Artikelnummer. 12406991 Leverandør Solveco Chemicals Leverandørnr. 1277

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Forhindrer mikrobiologisk kontaminering. Solveco Ethanol 70% +IPA Ph.Eur. giver en høj grad af desinfektion på glatte, synligt rene overflader. Det er effektivt mod bakterier, svampe og vira.

Fermented ethanol with a concentration of 70 vol - %, denatured with isopropanol. Meets and is tested according to the quality standards of the European Pharmacopoeia. Denatured Ethanol 70% is primarily used for disinfection and is highly effective on smooth, visibly clean surfaces. It is mainly used to prevent the spread of microbial infections. The product is effective against bacteria, fungi, and viruses.

Tekniske data

Kemisk navn eller materiale Ethyl Alcohol
CAS 12-34-5
Mængde 25 L
Emballage HDPE Bottle
Kogepunkt 78°C
Molekylvægt (g/mol) 46.069
Procent renhed 70%
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.