Få mere at vide
Ethanethiol-SD, CDN
Artikelnummer. 30321699
Skift visning
Mængde:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30321699 5 g
30328153 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321699 Leverandør CDN Leverandørnr. CDND56655G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ethanethiol-SD
CAS 33458-36-5
Molekylær formel C2 D H5 S
Mængde 5 g
Synonym Ethanethiol-d (8 CI, 9 CI), Ethanethiol-SD, Ethyl mercaptan-d, Thioethanol-d
InChI formel InChI=1 S/C2H6S/c1-2-3/h3H,2H2,1H3/i/hD
SMIL [2 H]SCC
IUPAC navn deuteriosulfanylethane
Molekylvægt (g/mol) 63.1402
Formel vægt 63.0253 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity