Få mere at vide
Ethametsulfuron-methyl, TRC
Artikelnummer. 30136869
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136869 500 mg
30004750 50 mg
30004768 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136869 Leverandør Toronto Research Chemicals Leverandørnr. TRCE675810500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 97780-06-8
Molekylær formel C15 H18 N6 O6 S
Molekylvægt (g/mol) 410.4
Synonym DPX-A 7881, Ethametsulfuron methyl ester, Ethametsulfuron-methyl, Muster, Salsa, Salsa 75WG, Benzoic acid, 2-[[[[[4-ethoxy-6-(methylamino)-1,3,5-triazin-2-yl]amino]carbonyl]amino]sulfonyl]-, methyl ester
IUPAC navn methyl 2-[[4-ethoxy-6-(methylamino)-1,3,5-triazin-2-yl]carbamoylsulfamoyl]benzoate
SMIL CCOc1nc(NC)nc(NC(=O)NS(=O)(=O)c2ccccc2C(=O)OC)n1
Tekniske data
Kemisk navn eller materiale Ethametsulfuron-methyl
Mængde 500 mg
InChI formel InChI=1S/C15H18N6O6S/c1-4-27-15-19-12(16-2)17-13(20-15)18-14(23)21-28(24,25)10-8-6-5-7-9(10)11(22)26-3/h5-8H,4H2,1-3H3,(H3,16,17,18,19,20,21,23)
Formel vægt 410.1009