Learn More
VITLAB™ ETFE Griffin Beaker
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
250 mL
400 mL
500 mL
600 mL
1000 mL
Description
- Griffin beakers made from ETFE (ethylene tetrafluoroethylene) with a black imprinted scale
- Transparent, with an easily readable black printed scale
- Very good chemical resistance
- Thermal stability from -100°C to +150°C
- Available in eight different sizes from 25 to 1000mL
Specifications
Color | Transparent |
Capacity (Metric) | 25 mL |
Material | ETFE |
For Use With (Application) | For Volume Measurement |
Height (Metric) | 50 mm |
Type | Griffin Beaker |
Autoclavable | Autoclavable |
Graduated | Yes |
Scale | Black Printed Scale |
Style | Griffin |