Learn More
Cytiva ETFE Capillary Tubing
Product Code. 11525734 Shop All Cytiva Products
Change view
Click to view available options
Overall Length (Metric):
2 m
3 m
Diameter (Metric) Inner:
0.75 mm
0.8 mm
1 mm
Unit Size:
Each
Pack of 2
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Overall Length (Metric) Diameter (Metric) Inner Unit Size
11525734 2 m 0.75 mm Pack of 2
10794662 2 m 0.8 mm Each
10722667 3 m 1 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11525734 Supplier Cytiva Supplier No. 18111974
  • ONLINE EXCLUSIVE PRICE
    525.00 DKK
    479.00 DKK / Pakke med 2
    Spare 46.00 DKK
    8% Slukket
På lager
For use with ÄKTA chromatography systems. Cytiva ETFE Capilary Tubing is available in a range of sizes.
Ethylenetetra fluoroethylene (ETFE) tubing for the controlled delivery of liquid in chromatography systems.
TRUSTED_SUSTAINABILITY
Specifications
For Use With ÄKTA Chromatography Systems
Type Capillary Tubing
Diameter (English) Outer 1/16 in.
Diameter (Metric) Inner 0.75 mm
Diameter (Metric) Outer 1.59 mm
Overall Length (Metric) 2 m
Material ETFE