Få mere at vide
Etanidazole, TRC
Artikelnummer. 30377965
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30377965 25 mg
30371808 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30377965 Leverandør Toronto Research Chemicals Leverandørnr. TRCE88923025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 22668-01-5
Molekylær formel C7 H10 N4 O4
Molekylvægt (g/mol) 214.18
Synonym N-(2-Hydroxyethyl)-2-nitro-1H-imidazole-1-acetamide
IUPAC navn N-(2-hydroxyethyl)-2-(2-nitroimidazol-1-yl)acetamide
SMIL OCCNC(=O)Cn1ccnc1[N+](=O)[O-]
Tekniske data
Kemisk navn eller materiale Etanidazole
Mængde 25 mg
InChI formel InChI=1S/C7H10N4O4/c12-4-2-8-6(13)5-10-3-1-9-7(10)11(14)15/h1,3,12H,2,4-5H2,(H,8,13)
Formel vægt 214.0702