Få mere at vide
Etafedrine Hydrochloride, LoGiCal
Artikelnummer. 30460011
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30460011 10 mg
1 options
Artikelnummer. 30460011 Leverandør LoGiCal Leverandørnr. LGCFOR0718.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Etafedrine Hydrochloride
CAS 530-35-8
Analyt- eller komponentnavne Etafedrine Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C12 H19 N O . Cl H
Certificeringer/compliance ISO 9001
Synonym (-)-Ethylephedrine hydrochloride, 2-Methylethylamino-1-phenyl-1-propanol hydrochloride, Menetyl, Nethamine, Nethamine Hydrochloride, Novedrin, Novedrin Hydrochloride, l-N-Ethylephedrine hydrochloride, Benzenemethanol, alpha-[1-(ethylmethylamino)ethyl]-, hydrochloride, [R-(R*,S*)]- (9CI)
InChI formel InChI=1S/C12H19NO.ClH/c1-4-13(3)10(2)12(14)11-8-6-5-7-9-11;/h5-10,12,14H,4H2,1-3H3;1H/t10-,12-;/m0./s1
Løsningstype Neat
SMIL Cl.CCN(C)[C@@H](C)[C@H](O)c1ccccc1
IUPAC navn (1R,2S)-2-[ethyl(methyl)amino]-1-phenylpropan-1-ol;hydrochloride
Industry Type Clinical Testing
Molekylvægt (g/mol) 229.75
Mængde 10 mg
Formel vægt 229.1233 g/mol