Få mere at vide
Estrone-2,4-d2, TRC
Artikelnummer. 30372530
Skift visning
Mængde:
0.5 mg
Artikelnummer. Mængde
30372530 0.5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30372530 Leverandør Toronto Research Chemicals Leverandørnr. TRCE8890610.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Estrone-2,4-D2
CAS 350820-16-5
Molekylær formel C18 2H2 H20 O2
Mængde 0.5 mg
Synonym Estra-1,3,5(10)-trien-17-one-2,4-d2, 3-hydroxy-, Estrone-(2,4-D2), 2,4-Dideutero-3-hydroxyestra-1,3,5(10)-trien-17-one, Estrone-2,4-D2, Estrone-d2
InChI formel InChI=1S/C18H22O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h3,5,10,14-16,19H,2,4,6-9H2,1H3/t14-,15-,16+,18+/m1/s1/i3D,10D
SMIL [2H]c1cc2[C@H]3CC[C@@]4(C)[C@@H](CCC4=O)[C@@H]3CCc2c([2H])c1O
IUPAC navn (8R,9S,13S,14S)-2,4-dideuterio-3-hydroxy-13-methyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-one
Molekylvægt (g/mol) 272.38
Formel vægt 272.1745