Få mere at vide
Estrone-2,4-d2, CDN
Artikelnummer. 30311457
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30311457 0.01 g
30314255 0.05 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30311457 Leverandør CDN Leverandørnr. CDND50050.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Estrone-2,4-D2
CAS 350820-16-5
Molekylær formel C18 2H2 H20 O2
Mængde 0.01 g
Synonym Estra-1,3,5(10)-trien-17-one-2,4-d2, 3-hydroxy-, Estrone-(2,4-D2), 2,4-Dideutero-3-hydroxyestra-1,3,5(10)-trien-17-one, Estrone-2,4-D2, Estrone-d2
InChI formel InChI=1 S/C18H22O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h3,5,10,14-16,19 H,2,4,6-9H2,1H3/t14-,15-,16+,18+/m1/s1/i3D,10 D
SMIL [2 H]c1cc2[C@H]3 CC[C@@]4(C)[C@@H](CCC4=O)[C@@H]3 CCc2c([2 H])c1O
IUPAC navn (8 R,9 S,13 S,14 S)-2,4-dideuterio-3-hydroxy-13-methyl-7,8,9,11,12,14,15,16-octahydro-6 H-cyclopenta[a]phenanthren-17-one
Molekylvægt (g/mol) 272.38
Formel vægt 272.1745 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity