Få mere at vide
Estrone-2,4,16,16-d4, CDN
Artikelnummer. 30315002
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30315002 0.05 g
30308835 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30315002 Leverandør CDN Leverandørnr. CDND36500.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Estrone D4
CAS 53866-34-5
Molekylær formel C18 D4 H18 O2
Mængde 0.05 g
Synonym Estrone-D4, Estra-1,3,5(10)-trien-17-one-2,4,16,16-d4, 3-hydroxy-, 3-Hydroxyestra-1,3,5(10)-trien-17-one-2,4,16,16-d4, Estrone-2,4,16,16-d4, Estrone-(2,4,16,16-D4), Estrone-2,4,16,16-D4
InChI formel InChI=1 S/C18H22O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h3,5,10,14-16,19 H,2,4,6-9H2,1H3/t14-,15-,16+,18+/m1/s1/i3D,7D2,10 D
SMIL [2 H]c1cc2[C@H]3 CC[C@@]4(C)[C@@H](CC([2 H])([2 H])C4=O)[C@@H]3 CCc2c([2 H])c1O
IUPAC navn (8 R,9 S,13 S,14 S)-2,4,16,16-tetradeuterio-3-hydroxy-13-methyl-6,7,8,9,11,12,14,15-octahydrocyclopenta[a]phenanthren-17-one
Molekylvægt (g/mol) 274.391
Formel vægt 274.187 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity