Få mere at vide
Invitrogen™ Estrogen Receptor beta Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA1311
Beskrivelse
PA1-311 detects estrogen receptor (ER) beta from human, non-human primate, mouse, ovine and rat tissues. PA1-311 has been successfully used in Western blot, immunocytochemistry and gel shift procedures. By Western blot, this antibody detects an ~55-62 kDa protein representing ER beta from rat brain homogenate. PA1-311 efficiently supershifts rat ER beta/ER response element (vitellogenin xenopus A2 gene). The PA1-311 immunogen is a synthetic peptide corresponding to residues A(55) E P Q K S P W C E A R S L E H(70) of rat ER beta. Recombinant ER beta expressed in baculovirus system (Cat. # RP-311 and RP-312) can be purchased for control experiments in Western blot and gel shift assays. PA1-311 can be used with blocking peptide PEP-011.
Estrogen Receptor beta (ER beta, NR3A2) protein is approximately 55kD, though multiple isoforms ranging from 36-59kD have been described. Human ER beta was initially cloned and characterized from testis. Estrogen Receptors (ER) are members of the steroid/thyroid hormone receptor superfamily of nuclear receptors. The estrogen receptor is a ligand-activated transcription factor, that when bound to estrogen hormone, induces a conformational change that allows dimerization and binding to estrogen response elements (ERE) in DNA. When bound to EREs, ER can positively or negatively regulate gene transcription through the recruitment of coactivator or corepressor proteins. There are two different forms of the estrogen receptor, alpha and beta, encoded by separate genes (ESR1 and ESR2, respectively). Estrogen receptor beta (ER beta) binds estrogens with an affinity similar to that of estrogen receptor alpha. Some ER beta isoforms dominantly inhibit the activity of estrogen receptor alpha in reproductive tissues. ER beta is found widely in many tissues throughout the body and can act as a potent tumor suppressor, playing a crucial role in many cancer types such as prostate cancer, autism spectrum disorder, medullary thyroid carcinoma, De Quervain Disease and risk of cardiovascular disease.
Tekniske data
| Estrogen Receptor beta | |
| Polyclonal | |
| Unconjugated | |
| ESR2 | |
| ER; ER beta; ER[b]; Erb; Erb2; Erbeta; ER-beta; Esr2; ESRB; ESR-BETA; ESTRB; estrogen receptor 2; estrogen receptor 2 (beta); estrogen receptor 2 (ER beta); estrogen receptor 2 beta; estrogen receptor beta; estrogen receptor beta 1 isoform deletion variant; estrogen receptor beta 4; NR3A2; Nuclear receptor subfamily 3 group A member 2; oestrogen receptor beta; steroid receptor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 13983, 2100, 25149, 443066 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Gel Shift, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 1mg/mL BSA and 0.05% sodium azide | |
| O08537, Q62986, Q92731, Q9TU15 | |
| ESR2 | |
| Synthetic peptide corresponding to residues A(55) E P Q K S P W C E A R S L E H(70) of rat ER beta. | |
| 50 μg | |
| Primary | |
| Human, Mouse, Monkey, Sheep, Rat | |
| Antibody | |
| IgG |
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.