missing translation for 'onlineSavingsMsg'
Få mere at vide

Estriol Triacetate, Mikromol™

Artikelnummer. 30049894
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30049894 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30049894 Leverandør Mikromol Leverandørnr. MM0130.150025

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Estriol Triacetate
TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Estriol Triacetate
CAS 2284-32-4
Analyt- eller komponentnavne Estriol Triacetate
Forsendelsestilstand Room Temperature
Molekylær formel C24 H30 O6
Certificeringer/compliance ISO 9001
Synonym Estriol Triacetate
InChI formel InChI=1S/C24H30O6/c1-13(25)28-17-6-8-18-16(11-17)5-7-20-19(18)9-10-24(4)21(20)12-22(29-14(2)26)23(24)30-15(3)27/h6,8,11,19-23H,5,7,9-10,12H2,1-4H3/t19-,20-,21+,22-,23+,24+/m1/s1
SMIL CC(=O)O[C@@H]1C[C@H]2[C@@H]3CCc4cc(OC(=O)C)ccc4[C@H]3CC[C@]2(C)[C@H]1OC(=O)C
IUPAC navn [(8R,9S,13S,14S,16R,17R)-16,17-diacetyloxy-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-3-yl] acetate
Molekylvægt (g/mol) 414.49
Mængde 25 mg
Formel vægt 414.2042 g/mol
Vis mere Vis mindre
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.