Få mere at vide
Estriol-d1, TRC
Artikelnummer. 30378653
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30378653 1 mg
30371798 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30378653 Leverandør Toronto Research Chemicals Leverandørnr. TRCE8889631MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 55727-98-5
Molekylær formel C18H23DO3
Molekylvægt (g/mol) 289.18
Synonym (16alpha,17beta)-Estra-1,3,5[10]-triene-3,16,17-triol-d1, Acifemine-d1, Colpogyn-d1, Destriol-d1, Ovesterin-d1, Ovestin-d1, Tridestrin-d1, Tricovex-d1
IUPAC navn (8R,9S,13S,14S,16R,17R)-17-deuterio-13-methyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthrene-3,16,17-triol
SMIL [2H][C@]1(O)[C@H](O)C[C@H]2[C@@H]3CCc4cc(O)ccc4[C@H]3CC[C@]12C
Tekniske data
Kemisk navn eller materiale Estriol-d1
Mængde 1 mg
InChI formel InChI=1S/C18H24O3/c1-18-7-6-13-12-5-3-11(19)8-10(12)2-4-14(13)15(18)9-16(20)17(18)21/h3,5,8,13-17,19-21H,2,4,6-7,9H2,1H3/t13-,14-,15+,16-,17+,18+/m1/s1/i17D
Formel vægt 289.1788