Få mere at vide
Estradiol EP Impurity G, Mikromol™
Artikelnummer. 30051360
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051360 100 mg
30049477 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30051360 Leverandør Mikromol Leverandørnr. MM0301.08
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Estradiol EP Impurity G
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 17-Oxoestra-1,3,5(10)-trien-3-yl Benzoate (Estrone Benzoate)
CAS 2393-53-5
Analyt- eller komponentnavne 17-Oxoestra-1,3,5(10)-trien-3-yl Benzoate (Estrone Benzoate)
Forsendelsestilstand Room Temperature
Molekylær formel C25 H26 O3
Certificeringer/compliance ISO 17025
Synonym Estradiol Benzoate Imp. G (EP), 17-Oxoestra-1,3,5(10)-trien-3-yl Benzoate, Estrone Benzoate, Estradiol Benzoate Impurity G
InChI formel InChI=1S/C25H26O3/c1-25-14-13-20-19-10-8-18(28-24(27)16-5-3-2-4-6-16)15-17(19)7-9-21(20)22(25)11-12-23(25)26/h2-6,8,10,15,20-22H,7,9,11-14H2,1H3/t20-,21-,22+,25+/m1/s1
SMIL C[C@]12CC[C@H]3[C@@H](CCc4cc(OC(=O)c5ccccc5)ccc34)[C@@H]1CCC2=O
IUPAC navn [(8R,9S,13S,14S)-13-methyl-17-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-3-yl] benzoate
Molekylvægt (g/mol) 374.47
Mængde 100 mg
Formel vægt 374.1882 g/mol