Få mere at vide
Estradiol Benzoate, LoGiCal
Specifications
Specifications
| Kemisk navn eller materiale | Estradiol Benzoate |
| CAS | 50-50-0 |
| Analyt- eller komponentnavne | Estradiol Benzoate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C25 H28 O3 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Estradiol Benzoate, Estradiol 3-benzoate, Agofollin, Agofollin Depot, Benovocylin, Benzhormovarine, Benzo-Gynoestryl, Benzoestrofol, Benzofoline, Benztrone, Cidirol, De Graafina, Diffollisterol, Difolliculine, Dimenformon benzoate, Diogyn B, Eston B, Estradiol benzoate, Estradiol monobenzoate, Estrogin, Femestrone, Folone, Graafina, Gynecormone, Gynformone, Hidroestron, Hormogynon, Mesalin, NSC 9566, Oestroform [BDH], Ovahormon benzoate, Ovasterol B, Ovocyclin Benzoate, Ovocyclin M, Ovocyclin MB, Pelanin benzoate, Primogyn B, Primogyn B oleosum, Progynon B, Progynon benzoate, Recthormone Oestradiol, Solestro, Unistradiol, (17beta)-Estra-1,3,5(10)-triene-3,17-diol 3-benzoate, 1,3,5(10)-Estratriene-3,17beta-diol 3-benzoate, 17beta-Estradiol 3-benzoate, 17beta-Estradiol benzoate, 3-Benzoyloxy-17beta-hydroxyestra-1,3,5(10)-triene, beta-Estradiol 3-benzoate, beta-Estradiol benzoate |
| InChI formel | InChI=1S/C25H28O3/c1-25-14-13-20-19-10-8-18(28-24(27)16-5-3-2-4-6-16)15-17(19)7-9-21(20)22(25)11-12-23(25)26/h2-6,8,10,15,20-23,26H,7,9,11-14H2,1H3/t20-,21-,22+,23+,25+/m1/s1 |
| Løsningstype | Neat |
| SMIL | C[C@]12CC[C@H]3[C@@H](CCc4cc(OC(=O)c5ccccc5)ccc34)[C@@H]1CC[C@@H]2O |
| Show More |