Få mere at vide
Estradiol 17-Valerate-d9, TRC
Artikelnummer. 30370373
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30370373 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30370373 Leverandør Toronto Research Chemicals Leverandørnr. TRCE88886210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Estradiol 17-Valerate-d9
CAS 1316648-98-2
Molekylær formel C23H23D9O3
Mængde 10 mg
Synonym (17β)-Estra-1,3,5(10)-triene-3,17-diol 17-Pentanoate-d9, Estradiol Valerate-d9, 3-Hydroxy-17β-(valeroyl-d9)oxyestra-1,3,5(10)-triene, Atladiol-d9, Climaval-d9, Deladiol, -d9 Delestrogen-d9, Delestrogen 4x-d9, Estradiol 17β-Valerate-d9, Estradiol Valerianate-d9, Estraval-d9, Femogex-d9, Gynokadin-d9, NSC 17590-d9, Neofollin-d9, Nuvelle-d9, Progynova-d9, Valergen-d9
InChI formel InChI=1S/C23H32O3/c1-3-4-5-22(25)26-21-11-10-20-19-8-6-15-14-16(24)7-9-17(15)18(19)12-13-23(20,21)2/h7,9,14,18-21,24H,3-6,8,10-13H2,1-2H3/t18-,19-,20+,21+,23+/m1/s1/i1D3,3D2,4D2,5D2
SMIL OC1=CC(CC[C@@]([H])([C@@]23[H])[C@]4([H])CC[C@@]2([C@@H](OC(C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])=O)CC3)C)=C4C=C1
Molekylvægt (g/mol) 365.55
Formel vægt 365.2916