missing translation for 'onlineSavingsMsg'
Learn More

Estradiol 17-Acetate, Mikromol™

Product Code. 18041720
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
18041720 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 18041720 missing translation for 'mfr' Mikromol missing translation for 'supplierNo' MM0300.110025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Estradiol 17-Acetate
TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 17-beta-Estradiol 17-acetate
CAS 1743-60-8
Analyt- eller komponentnavne 17-beta-Estradiol 17-acetate
Forsendelsestilstand Room Temperature
Molekylær formel C20 H26 O3
Certificeringer/compliance ISO 17025
Synonym Estra-1,3,5(10)-triene-3,17-diol (17β)-, 17-acetate, Estradiol, 17-acetate (6CI,7CI,8CI), 17β-Acetoxyestra-1,3,5(10)-trien-3-ol, 17β-Acetylestradiol, 17β-Estradiol 17-acetate, Estra-1,3,5(10)-triene-3,17β-diol 17-acetate, Estradiol 17-monoacetate, β-Estradiol 17-acetate
InChI formel InChI=1S/C20H26O3/c1-12(21)23-19-8-7-18-17-5-3-13-11-14(22)4-6-15(13)16(17)9-10-20(18,19)2/h4,6,11,16-19,22H,3,5,7-10H2,1-2H3/t16-,17-,18+,19+,20+/m1/s1
SMIL CC(=O)O[C@H]1CC[C@H]2[C@@H]3CCc4cc(O)ccc4[C@H]3CC[C@]12C
IUPAC navn [(8R,9S,13S,14S,17S)-3-hydroxy-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-17-yl] acetate
Molekylvægt (g/mol) 314.42
Mængde 25 mg
Formel vægt 314.1882 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.