missing translation for 'onlineSavingsMsg'
Få mere at vide

Esomeprazole magnesium, MedChemExpress

Artikelnummer. 16484597
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16484597 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16484597 Leverandør Medchem Express Leverandørnr. HYB1446

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress Esomeprazole magnesium ((S)-Omeprazole magnesium) is a potent and orally active H+, K+-ATPase inhibitor. Esomeprazole magnesium has the potential for upper intestinal disorders and gastroesophageal reflux disease research. Esomeprazole magnesium acts as an exosome inhibitor by blocking the exosome release via the inhibition of V-H+-ATPases.

Esomeprazole magnesium ((S)-Omeprazole magnesium) is a potent and orally active H+, K+-ATPase inhibitor. Esomeprazole magnesium has the potential for upper intestinal disorders and gastroesophageal reflux disease research. Esomeprazole magnesium acts as an exosome inhibitor by blocking the exosome release via the inhibition of V-H+-ATPases.

Tekniske data

Kemisk navn eller materiale Esomeprazole magnesium
CAS 161973-10-0
Farve Light Yellow
Renhedsgrad noter Research
Molekylær formel C34H36MgN6O6S2
Mængde 50 mg
Synonym (S)-Omeprazole magnesium (-)-Omeprazole magnesium
Opløselighedsinformation DMSO : ≥ 125 mg/mL (175.29 mM) ∣H2O : 3 mg/mL (4.21 mM; Need ultrasonic)
SMIL CC1=C(N=CC(C)=C1OC)CS2=O[Mg+2]3([N](C4=CC=C(OC)C=C4[N-]5)=C5S(CC(N=CC(C)=C6OC)=C6C)=O3)[N]7=C2[N-]C8=CC(OC)=CC=C78
Molekylvægt (g/mol) 713.12
Formel vægt 713.12
Procent renhed 95.0%
Grad Research
Fysisk form Solid
Til brug med (applikation) Neuroscience-Neuromodulation
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.