Få mere at vide
Esmolol Isopropyl Amine, TRC
Artikelnummer. 30012333
Skift visning
Mængde:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30012333 100 mg
30009791 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30012333 Leverandør Toronto Research Chemicals Leverandørnr. TRCE668035100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 83356-59-6
Molekylær formel C18 H30 N2 O3
Molekylvægt (g/mol) 322.44
Synonym 4-[2-Hydroxy-3-[(1-methylethyl)amino]propoxy]-N-(1-methylethyl)-benzenepropanamide
IUPAC navn 3-[4-[2-hydroxy-3-(isopropylamino)propoxy]phenyl]-N-isopropyl-propanamide
SMIL CC(C)NCC(O)COc1ccc(CCC(=O)NC(C)C)cc1
Tekniske data
Kemisk navn eller materiale Esmolol Isopropyl Amine
Mængde 100 mg
InChI formel InChI=1S/C18H30N2O3/c1-13(2)19-11-16(21)12-23-17-8-5-15(6-9-17)7-10-18(22)20-14(3)4/h5-6,8-9,13-14,16,19,21H,7,10-12H2,1-4H3,(H,20,22)
Formel vægt 322.2256