Få mere at vide
Eserine, SPEX CertiPrep™
Artikelnummer. 12939344 Shop alle SPEX Certiprep produkter
Skift visning
Mængde:
1 mL
Pakningsstørrelse:
1ml
Artikelnummer. Mængde unitSize
12939344 1 mL 1ml
1 options
Artikelnummer. 12939344 Leverandør SPEX Certiprep Leverandørnr. S3102
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
AnalyteCAS No.Concentration
Eserine57-47-61000μg/mL
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Kemisk navn eller materiale Eserine
Emballage 2mL Amber Prescored Glass Ampule
CAS 57-47-6, 67-56-1
CAS min % 0.1, 99.9
Koncentration eller sammensætningsnoter Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
CAS Max % 0.1, 99.9
Molekylær formel C15H21N3O2
Til brug med (udstyr) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
InChI nøgle PIJVFDBKTWXHHD-HIFRSBDPSA-N
SMIL CC12CCN(C1N(C3=C2C=C(C=C3)OC(=O)NC)C)C
IUPAC navn [(3aR,8bS)-3,4,8b-trimethyl-2,3a-dihydro-1H-pyrrolo[2,3-b]indol-7-yl] N-methylcarbamate
Opløsningsmiddel eller Matrix Methanol
Molekylvægt (g/mol) 275.352
PubChem CID 5983
Mængde 1 mL
ChEBI CHEBI:27953
Show More Show Less
Product intended for laboratory use only