missing translation for 'onlineSavingsMsg'
Få mere at vide

Eserethol, TRC

Artikelnummer. 30373656
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
2.5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30373656 10 mg
30369011 1 mg
30370687 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30373656 Supplier Toronto Research Chemicals Supplier No. TRCE14622010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 469-23-8
Molekylær formel C15H22N2O
Molekylvægt (g/mol) 246.35
Synonym 3aS,8aR)-5-Ethoxy-1,3a,8-trimethyl-1,2,3,3a,8,8a-hexahydropyrrolo[2,3-b]indole, Pyrrolo[2,3-b]indole, 5-ethoxy-1,2,3,3a,8,8a-hexahydro-1,3a,8-trimethyl-, (3aS-cis)-(3aS,8aR)-5-Ethoxy-1,2,3,3a,8,8a-hexahydro-1,3a,8-trimethylpyrrolo[2,3-b]indole(-)-Eserethole
SMIL CCOc1ccc2N(C)[C@H]3N(C)CC[C@@]3(C)c2c1

Specifications

Kemisk navn eller materiale Eserethol
Mængde 10 mg
InChI formel InChI=1S/C15H22N2O/c1-5-18-11-6-7-13-12(10-11)15(2)8-9-16(3)14(15)17(13)4/h6-7,10,14H,5,8-9H2,1-4H3/t14-,15+/m1/s1
Formel vægt 246.1732
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.