Få mere at vide
Escitalopram N-Oxide, Mikromol™
Artikelnummer. 30050647
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30050647 100 mg
1 options
Artikelnummer. 30050647 Leverandør Mikromol Leverandørnr. MM1162.05
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Escitalopram N-Oxide
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Escitalopram N-Oxide
CAS 917482-45-2
Analyt- eller komponentnavne Escitalopram N-Oxide
Forsendelsestilstand Room Temperature
Molekylær formel C20 H21 F N2 O2
Certificeringer/compliance ISO 9001
Synonym (1S)-1-[3-(Dimethyloxidoamino)propyl]-1-(4-fluorophenyl)-1,3-dhydro-5-isobenzofurancarbonitrile, Escitalopram N-Oxide, (S)-Citalopram N-Oxide, Escitalopram Oxalate Impurity H as (S)-Enantiomer, Escitalopram Impurity H as (S)-Enantiomer
InChI formel InChI=1S/C20H21FN2O2/c1-23(2,24)11-3-10-20(17-5-7-18(21)8-6-17)19-9-4-15(13-22)12-16(19)14-25-20/h4-9,12H,3,10-11,14H2,1-2H3/t20-/m0/s1
SMIL C[N+](C)([O-])CCC[C@]1(OCc2cc(ccc12)C#N)c3ccc(F)cc3
IUPAC navn 3-[(1S)-5-cyano-1-(4-fluorophenyl)-3H-2-benzofuran-1-yl]-N,N-dimethylpropan-1-amine oxide
Molekylvægt (g/mol) 340.39
Mængde 100 mg
Formel vægt 340.1587 g/mol