Learn More
Thermo Scientific™ ES Series FMS Lab Refrigerators
Reduce risk of explosion. These lab refrigerators are ideal for storage of potentially flammable material in general laboratory locations with limited space.
Brand:  Thermo Scientific™ 288R-AXV-TS
Additional Details : Weight : 90.00000kg
 View more versions of this product
Product Code. 15254737
  • 12141.00 DKK / Each
Estimated Shipment
11-05-2021
Add to basket
Description
Description
Reduce risk of accidental explosion and associated sample damage. The new Thermo Scientific™ ES Series FMS Lab Refrigerators are ideal for storing potentially flammable material in general laboratory settings. Household and commercial freezers are not suitable for such material: they have components in their electrical and refrigeration systems that can trigger explosions of flammable air-vapor mixtures inside the unit. These freezers have been constructed without components that could trigger an explosion or fire of the materials inside.
  • Integrated controller
  • Digital display
  • High- and low-temperature alarms
  • Door ajar alarm
  • Remote alarm contacts
  • Standard door locks
  • Low energy consumption
  • Access ports
  • CE/ATEX marked
  • Reversible doors
  • Hydrocarbon refrigerants
  • HCFC and CFC free insulation
Ordering Information: Available in 50Hz only; not available in North America or other 60Hz regions.
Specifications
Specifications
Defrost
HCFC and CFC free insulation
Alarm
1°C to 11°C
288 L
230 V
European
Hydrocarbon
156.5 cm
64 cm
Automatic
Reversible
Hydrocarbon
5
FMS units are designed, evaluated and comply with the ATEX Directive 94/9/EC Annex VIII and bear the ATEX mark.
230 V 50 Hz
50 Hz
90 kg
44.5 cm
138.2 cm
48 cm
59.5 cm
Videos
SDS
Documents
FMS units are not designed for use in Class I, Zone 0 or Zone 1 applications which require an explosion-proof unit.