Få mere at vide
Erythromycin Glucoheptonate, TRC
Artikelnummer. 30379278
Skift visning
Mængde:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30379278 250 mg
30370258 100 mg
30371831 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30379278 Leverandør Toronto Research Chemicals Leverandørnr. TRCE649995250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 23067-13-2
Molekylær formel C37 H67 N O13 . C7 H14 O8
Molekylvægt (g/mol) 960.11
Synonym Erythromycin Glucoheptonate (7CI), Erythromycin Monoglucoheptonate (Salt), Erythromycin D-Glycero-D-gulo-heptonate (Salt), Glucoheptonic Acid, compd. with Erythromycin (1:1), Oxacyclotetradecane Erythromycin deriv., Erythromycin Gluceptate, Erythromycin Glucoheptonic Acid Salt, Ilotycin Gluceptate, Ilotycin Glucoheptonate
IUPAC navn (3R,4S,5S,6R,7R,9R,11R,12R,13S,14R)-6-[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-14-ethyl-7,12,13-trihydroxy-4-[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy-3,5,7,9,11,13-hexamethyl-oxacyclotetradecane-2,10-dione;(2R,3R,4S,5R,6R)-2,3,4,5,6,7-hexahydroxyheptanoic acid
SMIL CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]3O[C@H](C)C[C@@H]([C@H]3O)N(C)C)[C@](C)(O)C[C@@H](C)C(=O)[C@H](C)[C@@H](O)[C@]1(C)O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)[C@@H](O)C(=O)O
Tekniske data
Kemisk navn eller materiale Erythromycin Gluceptate
Mængde 250 mg
InChI formel InChI=1S/C37H67NO13.C7H14O8/c1-14-25-37(10,45)30(41)20(4)27(39)18(2)16-35(8,44)32(51-34-28(40)24(38(11)12)15-19(3)47-34)21(5)29(22(6)33(43)49-25)50-26-17-36(9,46-13)31(42)23(7)48-26;8-1-2(9)3(10)4(11)5(12)6(13)7(14)15/h18-26,28-32,34,40-42,44-45H,14-17H2,1-13H3;2-6,8-13H,1H2,(H,14,15)/t18-,19-,20+,21+,22-,23+,24+,25-,26+,28-,29+,30-,31+,32-,34+,35-,36-,37-;2-,3-,4+,5-,6-/m11/s1
Formel vægt 959.5301