Få mere at vide
Erythromycin A 9,11-Imino Ether, TRC
Artikelnummer. 30010785
Skift visning
Mængde:
1 mg
2.5 mg
10 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30010785 2.5 mg 2.5mg
30012336 10 mg -
30010800 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30010785 Leverandør Toronto Research Chemicals Leverandørnr. TRCE6500252.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 161193-44-8
Molekylær formel C37H66N2O12
Molekylvægt (g/mol) 730.93
Synonym [1R-(1R*,2R*,3R*,6R*,7S*,8S*,9R*,10R*,12R*,15R*)]-7-[(2,6-Dideoxy-3-C-methyl-3-O-methyl-α-L-ribo-hexopyranosyl)oxy]-3-ethyl-2,10-dihydroxy-2,6,8,10,12,15-hexamethyl-9-[[3,4,6-trideoxy-3-(dimethylamino)-β-D-xylo-hexopyranosyl]oxy]-4,16-dioxa-14-azabicyclo[11.2.1]hexadec-13-en-5-one, (1R,2R,3R,6R,7S,8S,9R,10R,12R,15R)-7-[(2,6-Dideoxy-3-C-methyl-3-O-methyl-α-L-ribo-hexopyranosyl)oxy]-3-ethyl-2,10-dihydroxy-2,6,8,10,12,15-hexamethyl-9-[[3,4,6-trideoxy-3-(dimethylamino)-β-D-xylo-hexopyranosyl]oxy]-4,,16-Dioxa-14-azabicyclo[11.2.1]hexadec-13-en-5-one
SMIL O[C@@]([C@@H](CC)OC([C@H](C)[C@@H](O[C@@H]1O[C@@H](C)[C@H](O)[C@@](OC)(C)C1)[C@H](C)[C@@H](OC2[C@H](O)[C@@H](N(C)C)C[C@@H](C)O2)[C@@](O)(C)C[C@H]3C)=O)(C)[C@@H]4OC3=N[C@@H]4C
Tekniske data
Kemisk navn eller materiale Erythromycin A 9,11-Imino Ether
Renhedsgrad noter HPLC
Mængde 2.5 mg
InChI formel InChI=1S/C37H66N2O12/c1-14-25-37(10,44)31-22(6)38-32(50-31)18(2)16-35(8,43)30(51-34-27(40)24(39(11)12)15-19(3)46-34)20(4)28(21(5)33(42)48-25)49-26-17-36(9,45-13)29(41)23(7)47-26/h18-31,34,40-41,43-44H,14-17H2,1-13H3/t18-,19-,20+,21-,22-,23+,24+,25-,26+,27-,28+,29+,30-,31-,34?,35-,36-,37-/m1/s1
Procent renhed >95%