missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ertumaxomab Human anti-Chemical, Invitrogen™
Human Recombinant Monoclonal Antibody
Brand: Invitrogen MA541824
This item is not returnable.
View return policy
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Ertumaxomab Recombinant Monoclonal antibody specifically detects Ertumaxomab in Chemical samples. It is validated for ELISA
Specifications
Ertumaxomab | |
Recombinant Monoclonal | |
Unconjugated | |
CD340; CD3E; ERBB2; Musmus G2a kappa; Rat sp.G2b lambda; TPBs03 | |
Human CD3E/ERBB2/CD340 | |
100 μg | |
Primary | |
Antibody | |
IgG2a κ, IgG2b λ |
ELISA | |
1 mg/mL | |
PBS with no preservative; pH 7.4 | |
Human | |
Protein A | |
RUO | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ertumaxomab Human anti-Chemical, Invitrogen™ > 100 μg; Unconjugated
Spot an opportunity for improvement?Share a Content Correction