Få mere at vide
Tocris Bioscience™ Erlotinib Hydrochloride
Beskrivelse
Erlotinib hydrochloride is a reversible, potent EGFR tyrosine kinase inhibitor (IC50 = 2 nM); it reduces EGFR autophosphorylation in intact tumor cells (IC50 = 20 nM). Erlotinib is selective for cell lines showing PC-9 and H3255 mutations (IC50 values are 7 and 12 nM, respectively, sparing wild-type and T790 mutant EGFR. Erlotinib shows antiproliferative effects in cancer cell lines in vitro and promotes differentiation of urothelial organoids. Orally bioavailable.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-(3-Ethynylphenyl)-6,7-bis(2-methoxyethoxy)-4-quinazolinamine hydrochloride |
| CAS | 183319-69-9 |
| Mængde | 50 mg |
| Mål | EGF Receptor Inhibitors |
| Molekylær formel | C22H23N3O4.HCl |
| Renhed | 0.98 |