Få mere at vide
Erismodegib, TRC
Artikelnummer. 30378000
Skift visning
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30378000 5 mg
30373548 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30378000 Leverandør Toronto Research Chemicals Leverandørnr. TRCE6152005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 956697-53-3
Molekylær formel C26 H26 F3 N3 O3
Molekylvægt (g/mol) 485.5
Synonym [1,1'-Biphenyl]-3-carboxamide, N-[6-[(2R,6S)-2,6-dimethyl-4-morpholinyl]-3-pyridinyl]-2-methyl-4'-(trifluoromethoxy)-, rel-, rel-N-[6-[(2R,6S)-2,6-Dimethyl-4-morpholinyl]-3-pyridinyl]-2-methyl-4'-(trifluoromethoxy)[1,1'-biphenyl]-3-carboxamide, LDE 225, NVP-LDE 225, Odomzo, Sonidegib
IUPAC navn N-[6-[(2S,6R)-2,6-dimethylmorpholin-4-yl]pyridin-3-yl]-2-methyl-3-[4-(trifluoromethoxy)phenyl]benzamide
SMIL C[C@@H]1CN(C[C@H](C)O1)c2ccc(NC(=O)c3cccc(c3C)c4ccc(OC(F)(F)F)cc4)cn2
Tekniske data
Kemisk navn eller materiale Erismodegib
Mængde 5 mg
InChI formel InChI=1S/C26H26F3N3O3/c1-16-14-32(15-17(2)34-16)24-12-9-20(13-30-24)31-25(33)23-6-4-5-22(18(23)3)19-7-10-21(11-8-19)35-26(27,28)29/h4-13,16-17H,14-15H2,1-3H3,(H,31,33)/t16-,17+
Formel vægt 485.1926