Få mere at vide
Eriodictyol, MedChemExpress
Artikelnummer. 16451468
Encompass_Preferred
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
16451468 5 mg
1 options
Artikelnummer. 16451468 Leverandør Medchem Express Leverandørnr. HYN0637
MedChemExpress Eriodictyol is a flavonoid isolated from the Chinese herb, with antioxidant and anti-inflammatory activity. Eriodictyol induces Nrf2 signaling pathway. Eriodictyol is also a potent influenza RNA-dependent RNA polymerase inhibitor with an IC50 of 18 nM.
Eriodictyol is a flavonoid isolated from the Chinese herb, with antioxidant and anti-inflammatory activity. Eriodictyol induces Nrf2 signaling pathway. Eriodictyol is also a potent influenza RNA-dependent RNA polymerase inhibitor with an IC50 of 18 nM.
Tekniske data
Kemisk navn eller materiale Eriodictyol
CAS 552-58-9
Farve White
Renhedsgrad noter Research
Molekylær formel C15H12O6
Mængde 5 mg
Synonym Huazhongilexone
Opløselighedsinformation DMSO : 125 mg/mL (433.65 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMIL O=C1C[C@@H](C2=CC=C(O)C(O)=C2)OC3=CC(O)=CC(O)=C13
Molekylvægt (g/mol) 288.25
Formel vægt 288.25
Procent renhed 98.36%
Grad Research
Fysisk form Powder
Til brug med (applikation) COVID-19-immunoregulation