Få mere at vide
Tocris Bioscience™ (+/-)-Eriodictyol
Beskrivelse
(±)-Eriodictyol is a potent TRPV1 antagonist. Inhibits capsaicin (Cat. No. 0462) -induced Ca2+ influx in synaptosomes. Exhibits antinociceptive activity in animal models without affecting body temperature. Also antioxidant; protects against H2O2-induced neurotoxicity in vitro by activation of Nrf2/ARE signaling. Modeling studies predict binding to ACE2 preventing its interaction with SARS-CoV-2 S protein. Naturally-derived flavonoid. Orally bioavailable.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 2-(3,4-Dihydroxyphenyl)-2,3-dihydro-5,7-dihydroxy-4 H-1-benzopyran-4-one |
| CAS | 4049-38-1 |
| Mængde | 5 mg |
| Mål | TRPV Inhibitors |
| Molekylær formel | C15H12O6 |
| Renhed | 0.98 |