missing translation for 'onlineSavingsMsg'
Learn More

Eriochrome Black T, Pure, C.I.14645, Metal Indicator, Fisher Chemical™

Brand:  Fisher Chemical E/0100/48

Additional Details : CAS Number : 1787-61-7 Weight : 0.10000kg


Quantity 100g

 View more versions of this product

Product Code. 10467060

Quantity Price
1 573.00 DKK / 100g
4 571.45 DKK / 100g
16 542.88 DKK / 100g
In stock, ready to ship: 3
Add to basket


Description

Description

Specifications

Chemical Identifiers

1787-61-7
462.388
KJEKTGOJOIUSCG-GXTSIBQPSA-N
(4Z)-4-[(1-hydroxynaphthalen-2-yl)hydrazinylidene]-7-nitro-3-oxonaphthalene-1-sulfonic acid;sodium
C20H13N3NaO7S
3935
87355429
C1=CC=C2C(=C1)C=CC(=C2O)NN=C3C4=C(C=C(C=C4)[N+](=O)[O-])C(=CC3=O)S(=O)(=O)O.[Na]

Specifications

1787-61-7
3935
C1=CC=C2C(=C1)C=CC(=C2O)NN=C3C4=C(C=C(C=C4)[N+](=O)[O-])C(=CC3=O)S(=O)(=O)O.[Na]
462.388
461.38g/mol
Glass Jar
3.7
C20H13N3NaO7S
KJEKTGOJOIUSCG-GXTSIBQPSA-N
(4Z)-4-[(1-hydroxynaphthalen-2-yl)hydrazinylidene]-7-nitro-3-oxonaphthalene-1-sulfonic acid;sodium
87355429
Powder Solid
Black
100g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Eriochrome Black T

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents