Learn More
Eriochrome Black T (C.I. 14645), Supelco™
Product Code. 15854340
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
Product Code. Quantity Unit Size
15854340 25 g 25g
1 options
MQ200 Graded - For Research and Non-regulated Industrial Applications
Product Code. 15854340 Supplier Supelco Supplier No. 1031700025
 Restricted Item
Forventet forsendelse: 18-11-2026
MQ200 Graded - For Research and Non-regulated Industrial Applications
Indicator for complexometry ACS,Reag. Ph Eur
TRUSTED_SUSTAINABILITY
Specifications
CAS 1787-61-7
MDL Number MFCD00003935
Synonym Eriochrome black T (C.I. 14645), Chrome black T, 2-Hydroxy-1-(1-hydroxy-2-naphthylazo)-6-nitronaphthalene-4-sulfonic acid sodium salt, Eriochrome black T (C.I. 14645)
Quantity 25 g
Molecular Weight (g/mol) 461.38
Grade Research
Chemical Name or Material Eriochrome Black T (C.I. 14645)