Få mere at vide
Erdosteine, TRC
Artikelnummer. 30011797
Skift visning
Mængde:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30011797 5 g
30012059 1 g
30006566 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30011797 Leverandør Toronto Research Chemicals Leverandørnr. TRCE5960505G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 84611-23-4
Molekylær formel C8 H11 N O4 S2
Molekylvægt (g/mol) 249.31
Synonym Erdosteine;[[2-Oxo-2-[(tetrahydro-2-oxo-3-thienyl)amino]ethyl]thio]acetic acid, Dithiosteine, Erdostin, RV 144, Secresolv
IUPAC navn 2-[2-oxo-2-[(2-oxothiolan-3-yl)amino]ethyl]sulfanylacetic acid
SMIL OC(=O)CSCC(=O)NC1CCSC1=O
Tekniske data
Kemisk navn eller materiale Erdosteine
Renhedsgrad noter HPLC
Mængde 5 g
InChI formel InChI=1S/C8H11NO4S2/c10-6(3-14-4-7(11)12)9-5-1-2-15-8(5)13/h5H,1-4H2,(H,9,10)(H,11,12)
Formel vægt 249.0129
Procent renhed >95%